Assay Details
Target Gene Details
Entrez Gene ID: | 57172 |
Gene Name: | calcium/calmodulin dependent protein kinase IG |
Gene Aliases: |
CLICK3, CLICKIII, VWS1, dJ272L16.1 |
Location: |
Chr.1:209583700-209613939 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CAMK1G | NM_020439.2 | NP_065172.1 | ||
XM_017001866.1 | XP_016857355.1 | |||
XM_017001867.1 | XP_016857356.1 | |||
AF428261.1 | AAL28100.1 | |||
AK095713.1 | ||||
AK289675.1 | ||||
BC032787.1 | AAH32787.1 | |||
BG715920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3575639 | Chr.1:209579620 - 209597199 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)