Assay Details
Target Gene Details
Entrez Gene ID: | 9447 |
Gene Name: | absent in melanoma 2 |
Gene Aliases: |
PYHIN4 |
Location: |
Chr.1:159059226-159132351 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AIM2 | NM_004833.1 | NP_004824.1 | ||
XM_005245616.4 | XP_005245673.1 | |||
XM_017002848.1 | XP_016858337.1 | |||
AF024714.1 | AAB81613.1 | |||
AK292042.1 | ||||
BC010940.1 | AAH10940.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |