Assay Details
Target Gene Details
Entrez Gene ID: | 391059 |
Gene Name: | ferric chelate reductase 1 |
Gene Aliases: |
SDFR2, SDR2 |
Location: |
Chr.1:99708699-99766630 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FRRS1 | NM_001013660.2 | NP_001013682.2 | ||
XM_005270861.3 | XP_005270918.1 | |||
XM_011541451.2 | XP_011539753.1 | |||
XM_011541452.1 | XP_011539754.1 | |||
XM_017001271.1 | XP_016856760.1 | |||
AK131302.1 | BAD18470.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1002853 | Chr.1:99682647 - 99924231 on Build GRCh38 | Gain | AGL PALMD FRRS1 |
esv24594 | Chr.1:99729334 - 99738907 on Build GRCh38 | Loss | FRRS1 |
nsv998891 | Chr.1:99479588 - 99843250 on Build GRCh38 | Loss | LOC101928270 PALMD FRRS1 |
nsv526047 | Chr.1:99684466 - 99980490 on Build GRCh38 | Gain | AGL PALMD SLC35A3 FRRS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |