Assay Details
Target Gene Details
Entrez Gene ID: | 1600 |
Gene Name: | DAB1, reelin adaptor protein |
Gene Aliases: |
- |
Location: |
Chr.1:56997906-58250539 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DAB1 | NM_021080.3 | NP_066566.3 | ||
AB210012.1 | ||||
AF071062.1 | AAC70068.1 | |||
AK095513.1 | ||||
AK313144.1 | ||||
BC067445.1 | AAH67445.1 | |||
BC067446.1 | AAH67446.1 | |||
BC067447.1 | AAH67447.1 | |||
BC112938.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv546305 | Chr.1:56997653 - 57350817 on Build GRCh38 | Gain | DAB1 LOC105378749 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |