Assay Details
Target Gene Details
Entrez Gene ID: | 81607 |
Gene Name: | nectin cell adhesion molecule 4 |
Gene Aliases: |
EDSS1, LNIR, PRR4, PVRL4, nectin-4 |
Location: |
Chr.1:161070991-161089595 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NECTIN4 | NM_030916.2 | NP_112178.2 | ||
XM_005245508.3 | XP_005245565.1 | |||
XM_011510021.2 | XP_011508323.1 | |||
XM_011510022.2 | XP_011508324.1 | |||
XM_011510023.1 | XP_011508325.1 | |||
AB105191.1 | ||||
AB755430.1 | ||||
AF160477.1 | AAF82399.1 | |||
AF426163.1 | AAL23958.1 | |||
AK027753.1 | BAB55344.1 | |||
BC010423.1 | AAH10423.1 | |||
DB223835.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3351730 | Chr.1:161086938 - 161088736 on Build GRCh38 | Duplication | NECTIN4 |
nsv1161729 | Chr.1:161075405 - 161104302 on Build GRCh38 | Duplication | PFDN2 KLHDC9 NECTIN4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |