Assay Details
Target Gene Details
Entrez Gene ID: | 412 |
Gene Name: | steroid sulfatase (microsomal), isozyme S |
Gene Aliases: |
ARSC, ARSC1, ASC, ES, SSDD, XLI |
Location: |
Chr.X:7147252-7354643 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STS | NM_001320750.1 | NP_001307679.1 | ||
NM_001320751.1 | NP_001307680.1 | |||
NM_001320752.1 | NP_001307681.1 | |||
AM072428.1 | ||||
AM072429.1 | ||||
AM282551.1 | ||||
AM282552.1 | ||||
DA327298.1 | ||||
DA406759.1 | ||||
FM209047.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3576734 | Chr.X:6537111 - 7202526 on Build GRCh38 | Gain | PUDP MIR4767 STS |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |