Assay Details
Target Gene Details
Entrez Gene ID: | 11238 |
Gene Name: | carbonic anhydrase 5B |
Gene Aliases: |
CA-VB |
Location: |
Chr.X:15738281-15787625 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CA5B | NM_007220.3 | NP_009151.1 | ||
XM_005274442.4 | XP_005274499.1 | |||
XM_017029247.1 | XP_016884736.1 | |||
XM_017029249.1 | XP_016884738.1 | |||
AB021660.1 | BAA76671.1 | |||
AK291050.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2739978 | Chr.X:15696314 - 15775807 on Build GRCh38 | Deletion | CA5BP1 CA5B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |