Assay Details
Target Gene Details
Entrez Gene ID: | 4281 |
Gene Name: | midline 1 |
Gene Aliases: |
BBBG1, FXY, GBBB1, MIDIN, OGS1, OS, OSX, RNF59, TRIM18, XPRF, ZNFXY |
Location: |
Chr.X:10445310-11111264 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MID1 | NM_000381.3 | 10 | 5360 | NP_000372.1 |
NM_001098624.2 | 10 | 5290 | NP_001092094.1 | |
NM_001193277.1 | 10 | 5149 | NP_001180206.1 | |
NM_033289.1 | 10 | 5246 | NP_150631.1 | |
NM_033290.3 | 10 | 5152 | NP_150632.1 | |
XM_005274536.1 | 11 | 5325 | XP_005274593.1 | |
XM_005274537.1 | 11 | 5302 | XP_005274594.1 | |
XM_006724492.3 | 11 | 5924 | XP_006724555.1 | |
XM_011545525.2 | 13 | 5626 | XP_011543827.1 | |
XM_011545526.2 | 13 | 5661 | XP_011543828.1 | |
XM_011545527.2 | 11 | 5571 | XP_011543829.1 | |
XM_017029534.1 | 11 | 5491 | XP_016885023.1 | |
XM_017029535.1 | 13 | 5512 | XP_016885024.1 | |
XM_017029536.1 | 12 | 5588 | XP_016885025.1 | |
XM_017029537.1 | 12 | 5359 | XP_016885026.1 | |
XM_017029538.1 | 10 | 5058 | XP_016885027.1 | |
XM_017029539.1 | 10 | 5220 | XP_016885028.1 | |
AF041210.1 | 9 | 4849 | AAC33002.1 | |
EF217425.1 | 1 | 2955 | ||
EF217426.1 | 1 | 2955 | ||
EF532594.1 | 1 | 466 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |