Assay Details
Target Gene Details
Entrez Gene ID: | 8228 |
Gene Name: | patatin like phospholipase domain containing 4 |
Gene Aliases: |
DXS1283E, GS2, iPLA2eta |
Location: |
Chr.X:7898763-7928587 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PNPLA4 | NM_001142389.1 | NP_001135861.1 | ||
NM_001172672.1 | NP_001166143.1 | |||
NM_004650.2 | NP_004641.1 | |||
XM_011545588.1 | XP_011543890.1 | |||
AK289888.1 | ||||
AK304586.1 | ||||
BC020746.1 | AAH20746.1 | |||
BM850121.1 | ||||
U03886.1 | AAA16491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2752335 | Chr.X:7862723 - 8469952 on Build GRCh38 | Gain | PNPLA4 VCX3B VCX2 MIR651 |
nsv470340 | Chr.X:7857284 - 8463464 on Build GRCh38 | Gain | PNPLA4 VCX2 MIR651 |
dgv499n21 | Chr.X:7777235 - 8383635 on Build GRCh38 | Gain | VCX PNPLA4 VCX2 MIR651 |
esv3576759 | Chr.X:7742804 - 8254647 on Build GRCh38 | Gain | VCX PNPLA4 VCX2 MIR651 |
esv2752336 | Chr.X:7904862 - 8063263 on Build GRCh38 | Loss | PNPLA4 |
esv3576758 | Chr.X:7596710 - 8167603 on Build GRCh38 | Gain | VCX PNPLA4 MIR651 |
esv2422194 | Chr.X:7481793 - 8034821 on Build GRCh38 | Duplication | VCX PNPLA4 |
nsv6786 | Chr.X:7886158 - 7916141 on Build GRCh38 | Insertion | PNPLA4 |
dgv1n30 | Chr.X:7824572 - 8147412 on Build GRCh38 | Gain | VCX PNPLA4 MIR651 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |