Assay Details
Target Gene Details
Entrez Gene ID: | 1741 |
Gene Name: | discs large MAGUK scaffold protein 3 |
Gene Aliases: |
MRX, MRX90, NEDLG, PPP1R82, SAP102, XLMR |
Location: |
Chr.X:70444850-70505490 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLG3 | NM_001166278.1 | NP_001159750.1 | ||
NM_020730.2 | NP_065781.1 | |||
NM_021120.3 | NP_066943.2 | |||
XM_005262248.3 | XP_005262305.1 | |||
XM_006724625.2 | XP_006724688.1 | |||
XM_006724626.2 | XP_006724689.1 | |||
XM_011530883.1 | XP_011529185.1 | |||
XM_017029322.1 | XP_016884811.1 | |||
XM_017029323.1 | XP_016884812.1 | |||
XM_017029324.1 | XP_016884813.1 | |||
XM_017029326.1 | XP_016884815.1 | |||
XM_017029327.1 | XP_016884816.1 | |||
AB033058.1 | ||||
AB209328.1 | BAD92565.1 | |||
AK303377.1 | ||||
AK304020.1 | ||||
AK315041.1 | ||||
AK316518.1 | ||||
BC093864.1 | AAH93864.1 | |||
BC093866.1 | AAH93866.1 | |||
U49089.1 | AAB61453.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss | SLC7A3 NONO SNX12 FOXO4 CXorf65 DLG3-AS1 LOC101059915 KIF4A ZMYM3 LOC100132741 CXorf49 INGX MED12 GDPD2 DLG3 ACRC CXCR3 TEX11 LOC100129291 LOC105373244 IL2RG OGT CXorf49B NLGN3 TAF1 GJB1 LINC00891 ITGB1BP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |