Assay Details
Target Gene Details
Entrez Gene ID: | 63035 |
Gene Name: | BCL6 corepressor-like 1 |
Gene Aliases: |
BCoR-L1, CXorf10 |
Location: |
Chr.X:129980285-130058083 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCORL1 | XM_005262452.4 | XP_005262509.2 | ||
XM_005262453.4 | XP_005262510.2 | |||
XM_005262454.3 | XP_005262511.2 | |||
XM_005262455.4 | XP_005262512.2 | |||
XM_005262456.4 | XP_005262513.2 | |||
XM_006724776.3 | XP_006724839.1 | |||
XM_006724777.3 | XP_006724840.1 | |||
XM_006724779.2 | XP_006724842.1 | |||
XM_017029721.1 | XP_016885210.1 | |||
BE244710.1 | ||||
BE246581.1 | ||||
DA227697.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528290 | Chr.X:129635682 - 130429852 on Build GRCh38 | Gain | ELF4 SASH3 GPR119 XPNPEP2 RAB33A ZNF280C BCORL1 AIFM1 RBMX2 SLC25A14 ZDHHC9 APLN UTP14A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |