Assay Details
Target Gene Details
Entrez Gene ID: | 2258 |
Gene Name: | fibroblast growth factor 13 |
Gene Aliases: |
FGF-13, FGF2, FHF-2, FHF2 |
Location: |
Chr.X:138631573-139222889 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FGF13 | NM_001139498.1 | NP_001132970.1 | ||
NM_001139500.1 | NP_001132972.1 | |||
NM_001139501.1 | NP_001132973.1 | |||
NM_001139502.1 | NP_001132974.1 | |||
NM_004114.3 | NP_004105.1 | |||
NM_033642.2 | NP_378668.1 | |||
XM_005262399.1 | XP_005262456.1 | |||
AF100143.1 | AAD16400.1 | |||
AF100144.1 | AAD16401.1 | |||
AK289960.1 | ||||
AK297545.1 | ||||
AK303685.1 | ||||
AK316170.1 | ||||
BC012347.1 | AAH12347.1 | |||
BC034340.1 | AAH34340.1 | |||
BU195176.1 | ||||
U66198.1 | AAB18914.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525867 | Chr.X:138498822 - 139357684 on Build GRCh38 | Gain | MIR504 FGF13 LINC00889 FGF13-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |