Assay Details
Target Gene Details
Entrez Gene ID: | 11326 |
Gene Name: | V-set and immunoglobulin domain containing 4 |
Gene Aliases: |
CRIg, Z39IG |
Location: |
Chr.X:66021738-66040125 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VSIG4 | NM_001100431.1 | NP_001093901.1 | ||
NM_001184830.1 | NP_001171759.1 | |||
NM_001184831.1 | NP_001171760.1 | |||
NM_001257403.1 | NP_001244332.1 | |||
NM_007268.2 | NP_009199.1 | |||
XM_017029251.1 | XP_016884740.1 | |||
AJ132502.1 | CAB51536.1 | |||
AK075153.1 | ||||
AK075171.1 | ||||
AK293215.1 | ||||
AK297050.1 | ||||
AK298389.1 | ||||
AK301417.1 | ||||
AL542949.3 | ||||
AL552873.3 | ||||
AL553835.3 | ||||
AY358341.1 | AAQ88707.1 | |||
BC010525.1 | AAH10525.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6933 | Chr.X:66003429 - 66036427 on Build GRCh38 | Insertion | MIR223 VSIG4 |
esv3576919 | Chr.X:65837678 - 66804698 on Build GRCh38 | Gain | EDA2R HEPH MIR223 VSIG4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |