Assay Details
Target Gene Details
Entrez Gene ID: | 57826 |
Gene Name: | RAP2C, member of RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.X:132203024-132219480 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAP2C | NM_001271186.1 | NP_001258115.1 | ||
NM_001271187.1 | NP_001258116.1 | |||
NM_021183.4 | NP_067006.3 | |||
NR_073150.1 | ||||
XM_006724775.2 | XP_006724838.1 | |||
XM_011531376.1 | XP_011529678.1 | |||
AK124801.1 | ||||
AK310997.1 | ||||
AL049685.2 | CAB41256.1 | |||
AL832461.1 | ||||
AY298955.1 | AAP55684.1 | |||
BC003403.1 | AAH03403.1 | |||
BI767215.1 | ||||
BX399437.2 | ||||
BX647223.1 | ||||
CN308919.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)