Assay Details
Target Gene Details
Entrez Gene ID: | 5988 |
Gene Name: | ret finger protein like 1 |
Gene Aliases: |
RNF78 |
Location: |
Chr.22:29403031-29442491 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RFPL1 | XM_011530298.2 | XP_011528600.1 | ||
XM_017028900.1 | XP_016884389.1 | |||
XM_017028901.1 | XP_016884390.1 | |||
XM_017028902.1 | XP_016884391.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059994 | Chr.22:29401384 - 29438777 on Build GRCh38 | Gain | RFPL1 RFPL1S |
nsv834171 | Chr.22:29230413 - 29433855 on Build GRCh38 | Loss | GAS2L1 RFPL1 EMID1 RASL10A RHBDD3 EWSR1 SNORD125 AP1B1 MIR3653 |
nsv1060197 | Chr.22:29421518 - 29446522 on Build GRCh38 | Gain | RFPL1 RFPL1S |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |