Assay Details
Target Gene Details
Entrez Gene ID: | 150223 |
Gene Name: | YdjC homolog (bacterial) |
Gene Aliases: |
- |
Location: |
Chr.22:21628089-21630326 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YDJC | NM_001017964.1 | NP_001017964.1 | ||
XM_011529906.2 | XP_011528208.1 | |||
BC110075.1 | AAI10076.1 | |||
BC133043.1 | ||||
BC133047.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160730 | Chr.22:21373849 - 21633730 on Build GRCh38 | Deletion | TMEM191C CCDC116 RIMBP3C PI4KAP2 HIC2 YDJC UBE2L3 RIMBP3B |
nsv516806 | Chr.22:21628603 - 21642781 on Build GRCh38 | Loss | CCDC116 YDJC SDF2L1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |