Assay Details
Target Gene Details
Entrez Gene ID: | 23786 |
Gene Name: | BCL2 like 13 |
Gene Aliases: |
BCL-RAMBO, Bcl2-L-13, MIL1 |
Location: |
Chr.22:17628855-17730855 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9871 | Chr.22:17545048 - 17683541 on Build GRCh38 | Loss | BCL2L13 SLC25A18 CECR2 LOC101929372 ATP6V1E1 |
nsv526826 | Chr.22:17411511 - 17836413 on Build GRCh38 | Gain | BCL2L13 LINC00528 SLC25A18 CECR2 LOC101929372 MICAL3 ATP6V1E1 MIR3198-1 BID |
esv3893429 | Chr.22:17600825 - 17678777 on Build GRCh38 | Gain | BCL2L13 ATP6V1E1 |
esv3893428 | Chr.22:17578563 - 18040448 on Build GRCh38 | Loss | BCL2L13 FLJ41941 LINC00528 SLC25A18 LOC101929372 MIR648 MICAL3 ATP6V1E1 MIR3198-1 BID |
dgv4465n100 | Chr.22:17577691 - 17706530 on Build GRCh38 | Gain | BCL2L13 SLC25A18 LOC101929372 ATP6V1E1 |
nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss | LOC100996415 SLC25A18 LOC102725072 MICAL3 PEX26 LOC729461 ATP6V1E1 TUBA8 BID LOC642643 BCL2L13 CECR2 LOC100996401 PI4KAP1 RIMBP3 FLJ41941 USP18 LOC100996432 FAM230A TMEM191B LINC00528 LOC101929372 MIR648 GGT3P MIR3198-1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73376764] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |