Assay Details
Target Gene Details
Entrez Gene ID: | 27124 |
Gene Name: | inositol polyphosphate-5-phosphatase J |
Gene Aliases: |
INPP5, PIB5PA, PIPP |
Location: |
Chr.22:31107736-31134697 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
INPP5J | XM_011530143.1 | XP_011528445.1 | ||
XM_017028771.1 | XP_016884260.1 | |||
XM_017028773.1 | XP_016884262.1 | |||
AK308921.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv964621 | Chr.22:31107865 - 31113534 on Build GRCh38 | Duplication |
![]() |
nsv829173 | Chr.22:31054057 - 31147970 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv9896 | Chr.22:30973218 - 31117947 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)