Assay Details
Target Gene Details
Entrez Gene ID: | 25817 |
Gene Name: | family with sequence similarity 19 member A5, C-C motif chemokine like |
Gene Aliases: |
QLLK5208, TAFA-5, TAFA5, UNQ5208 |
Location: |
Chr.22:48489460-48751935 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM19A5 | NM_001082967.2 | NP_001076436.1 | ||
AF131851.1 | AAD20061.1 | |||
BC039396.1 | AAH39396.2 | |||
DB481230.1 |
Target Gene Details
Entrez Gene ID: | 284933 |
Gene Name: | uncharacterized LOC284933 |
Gene Aliases: |
- |
Location: |
Chr.22:48538900-48547387 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC284933 | NR_038917.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv778n67 | Chr.22:48453824 - 48657701 on Build GRCh38 | Gain |
![]() ![]() |
nsv955186 | Chr.22:48484289 - 48570188 on Build GRCh38 | Deletion |
![]() ![]() |
nsv1064218 | Chr.22:48472061 - 48792098 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv834232 | Chr.22:48358817 - 48553110 on Build GRCh38 | Loss |
![]() ![]() |
nsv589147 | Chr.22:48310704 - 48549198 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)