Assay Details
Target Gene Details
Entrez Gene ID: | 23760 |
Gene Name: | phosphatidylinositol transfer protein beta |
Gene Aliases: |
PI-TP-beta, PtdInsTP, VIB1B |
Location: |
Chr.22:27851669-27919306 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PITPNB | NM_001284277.1 | NP_001271206.1 | ||
NM_001284278.1 | NP_001271207.1 | |||
NM_012399.4 | NP_036531.1 | |||
XM_011530052.2 | XP_011528354.1 | |||
XM_017028707.1 | XP_016884196.1 | |||
AK302367.1 | ||||
AK313064.1 | ||||
BC018704.1 | AAH18704.1 | |||
BC031427.1 | AAH31427.1 | |||
BU147254.1 | ||||
CR456541.1 | CAG30427.1 | |||
D30037.1 | BAA06277.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv588878 | Chr.22:27852852 - 27918125 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)