Assay Details
Target Gene Details
Entrez Gene ID: | 149998 |
Gene Name: | lipase I |
Gene Aliases: |
CT17, LPDL, PLA1C, PRED5, mPA-PLA1 beta |
Location: |
Chr.21:14108813-14210953 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPI | NM_001302998.1 | NP_001289927.1 | ||
NM_001302999.1 | NP_001289928.1 | |||
NM_001303000.1 | NP_001289929.1 | |||
NM_001303001.1 | NP_001289930.1 | |||
NM_198996.3 | NP_945347.2 | |||
AY197607.1 | AAP37476.1 | |||
JN387910.1 | ||||
JN387911.1 | ||||
JN387912.1 | ||||
JN387913.1 | ||||
JN387914.1 | ||||
JN387915.1 | ||||
JN387916.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3646422 | Chr.21:14095581 - 14339672 on Build GRCh38 | Gain | ABCC13 RBM11 LIPI |
nsv1056787 | Chr.21:14035183 - 14157970 on Build GRCh38 | Loss | LOC105377134 LIPI |
nsv1058202 | Chr.21:13334446 - 14256347 on Build GRCh38 | Loss | LOC105377134 MIR3118-1 LOC102724188 CYP4F29P ANKRD20A11P RBM11 MIR8069-2 MIR3156-3 POTED LIPI |
nsv587043 | Chr.21:14129201 - 14178114 on Build GRCh38 | Loss | LIPI |
nsv1058072 | Chr.21:14000188 - 14195850 on Build GRCh38 | Gain | LOC105377134 LIPI |
esv3893371 | Chr.21:14104569 - 14339246 on Build GRCh38 | Gain | ABCC13 RBM11 LIPI |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |