Assay Details
Target Gene Details
Entrez Gene ID: | 2551 |
Gene Name: | GA binding protein transcription factor alpha subunit |
Gene Aliases: |
E4TF1-60, E4TF1A, NFT2, NRF2, NRF2A, RCH04A07 |
Location: |
Chr.21:25734947-25772460 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GABPA | NM_001197297.1 | NP_001184226.1 | ||
NM_002040.3 | NP_002031.2 | |||
XM_005260938.4 | XP_005260995.1 | |||
XM_011529520.2 | XP_011527822.1 | |||
XM_011529521.2 | XP_011527823.1 | |||
XM_017028313.1 | XP_016883802.1 | |||
AK223164.1 | BAD96884.1 | |||
AK313545.1 | ||||
BC035031.2 | AAH35031.1 | |||
BX647755.1 | ||||
D13318.1 | BAA02575.1 | |||
EU159453.1 | ||||
JQ313900.1 | ||||
U13044.1 | AAA65706.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1064561 | Chr.21:25714853 - 25750046 on Build GRCh38 | Loss | JAM2 GABPA ATP5J |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |