Assay Details
Target Gene Details
Entrez Gene ID: | 54014 |
Gene Name: | bromodomain and WD repeat domain containing 1 |
Gene Aliases: |
C21orf107, DCAF19, N143, WDR9 |
Location: |
Chr.21:39184176-39321595 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BRWD1 | XM_017028373.1 | XP_016883862.1 |
Target Gene Details
Entrez Gene ID: | 100874093 |
Gene Name: | BRWD1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.21:39315707-39323218 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BRWD1-AS1 | NR_046655.1 | |||
DB057251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv587480 | Chr.21:39199320 - 39329243 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv587482 | Chr.21:39312501 - 39334668 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv587483 | Chr.21:39314401 - 39346235 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)