Assay Details
Target Gene Details
Entrez Gene ID: | 10694 |
Gene Name: | chaperonin containing TCP1 subunit 8 |
Gene Aliases: |
C21orf112, Cctq, D21S246, PRED71 |
Location: |
Chr.21:29056322-29073797 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCT8 | NM_001282907.1 | NP_001269836.1 | ||
NM_001282908.1 | NP_001269837.1 | |||
NM_001282909.1 | NP_001269838.1 | |||
NM_006585.3 | NP_006576.2 | |||
AK222490.1 | BAD96210.1 | |||
AK293705.1 | ||||
AK298801.1 | ||||
AK308063.1 | ||||
BC012584.1 | AAH12584.1 | |||
BC072001.1 | AAH72001.1 | |||
BC095470.1 | AAH95470.1 | |||
D13627.1 | ||||
DB301557.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |