Assay Details
Target Gene Details
Entrez Gene ID: | 26051 |
Gene Name: | protein phosphatase 1 regulatory subunit 16B |
Gene Aliases: |
ANKRD4, TIMAP |
Location: |
Chr.20:38805693-38923024 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP1R16B | NM_001172735.2 | NP_001166206.1 | ||
NM_015568.3 | NP_056383.1 | |||
XM_017027785.1 | XP_016883274.1 | |||
AB020630.2 | ||||
AK311244.1 | ||||
BC131801.1 | ||||
BC152467.1 | ||||
HY125679.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1058457 | Chr.20:38277492 - 38844429 on Build GRCh38 | Loss | LBP SNHG11 RALGAPB SNORA60 SLC32A1 LOC149684 BPI ARHGAP40 SNORA71E ADIG PPP1R16B SNORA71C ACTR5 SNORA71D SNORA71A SNORA71B MIR548O2 SNHG17 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |