Assay Details
Target Gene Details
Entrez Gene ID: | 58495 |
Gene Name: | ovo like zinc finger 2 |
Gene Aliases: |
EUROIMAGE566589, PPCD1, ZNF339 |
Location: |
Chr.20:18024152-18059188 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OVOL2 | NM_001303461.1 | NP_001290390.1 | ||
NM_001303462.1 | NP_001290391.1 | |||
NM_021220.3 | NP_067043.2 | |||
AK022284.1 | BAB14002.1 | |||
AL079276.1 | CAB45151.1 | |||
BC006148.1 | AAH06148.1 | |||
BE791524.1 | ||||
BM563561.1 | ||||
BT007295.1 | AAP35959.1 | |||
BX331154.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv585664 | Chr.20:18005414 - 18038630 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)