Assay Details
Target Gene Details
Entrez Gene ID: | 101926987 |
Gene Name: | DLGAP4 antisense RNA 1 |
Gene Aliases: |
CCAT7 |
Location: |
Chr.20:36507702-36573275 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLGAP4-AS1 | NR_109939.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833967 | Chr.20:36477427 - 36658869 on Build GRCh38 | Loss | DLGAP4-AS1 TGIF2 C20orf24 SLA2 NDRG3 TGIF2-C20orf24 DLGAP4 MYL9 |
nsv1057192 | Chr.20:36514051 - 36839310 on Build GRCh38 | Loss | DSN1 DLGAP4-AS1 TGIF2 C20orf24 SLA2 SOGA1 NDRG3 TGIF2-C20orf24 DLGAP4 MYL9 |