Assay Details
Target Gene Details
Entrez Gene ID: | 51225 |
Gene Name: | ABI family member 3 |
Gene Aliases: |
NESH, SSH3BP3 |
Location: |
Chr.17:49210227-49223225 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABI3 | NM_001135186.1 | NP_001128658.1 | ||
NM_016428.2 | NP_057512.1 | |||
XM_005257429.2 | XP_005257486.1 | |||
XM_011524873.1 | XP_011523175.1 | |||
XM_017024721.1 | XP_016880210.1 | |||
AB037886.1 | BAA90667.1 | |||
AL136709.1 | CAB66644.1 | |||
BC007780.2 | AAH07780.1 | |||
BI768790.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2074 | Chr.17:49208171 - 49233994 on Build GRCh38 | Insertion | GNGT2 PHOSPHO1 ABI3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |