Assay Details
Target Gene Details
Entrez Gene ID: | 5697 |
Gene Name: | peptide YY |
Gene Aliases: |
PYY-I, PYY1 |
Location: |
Chr.17:43952733-44024260 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PYY | NM_004160.5 | NP_004151.3 | ||
XM_011525035.1 | XP_011523337.1 | |||
BC041057.1 | AAH41057.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833454 | Chr.17:43940848 - 44158311 on Build GRCh38 | Loss | HDAC5 NAGS LOC105371789 PPY LSM12 G6PC3 TMEM101 PYY C17orf53 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |