Assay Details
Target Gene Details
Entrez Gene ID: | 6352 |
Gene Name: | C-C motif chemokine ligand 5 |
Gene Aliases: |
D17S136E, RANTES, SCYA5, SIS-delta, SISd, TCP228, eoCP |
Location: |
Chr.17:35871491-35880373 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL5 | NM_001278736.1 | NP_001265665.1 | ||
NM_002985.2 | NP_002976.2 | |||
AF043341.1 | AAC03541.1 | |||
AF266753.1 | AAF73070.1 | |||
AK312212.1 | ||||
BC008600.1 | AAH08600.1 | |||
CR980938.1 | ||||
DQ230537.1 | ||||
M21121.1 | AAA36725.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059795 | Chr.17:35828724 - 36157706 on Build GRCh38 | Gain | CCL4 CCL15-CCL14 LRRC37A8P CCL15 CCL23 LYZL6 TAF15 CCL5 CCL3 CCL18 LOC101927339 CCL14 HEATR9 RDM1 CCL16 |
esv3892999 | Chr.17:35252863 - 35925533 on Build GRCh38 | Loss | LOC105371933 SLFN12L LRRC37A8P MMP28 SLFN13 GAS2L2 SNORD7 TAF15 CCL5 SLFN11 HEATR9 RASL10B C17orf50 SLFN14 RDM1 SLFN5 PEX12 LOC105371743 SLFN12 AP2B1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |