Assay Details
Target Gene Details
Entrez Gene ID: | 26207 |
Gene Name: | phosphatidylinositol transfer protein, cytoplasmic 1 |
Gene Aliases: |
M-RDGB-beta, MRDGBbeta, RDGB-BETA, RDGBB, RDGBB1 |
Location: |
Chr.17:67377281-67697263 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PITPNC1 | NM_012417.3 | NP_036549.2 | ||
NM_181671.2 | NP_858057.1 | |||
XM_005257216.2 | XP_005257273.1 | |||
XM_017024444.1 | XP_016879933.1 | |||
AF171102.1 | AAF06148.1 | |||
AK290838.1 | ||||
BC007905.2 | AAH07905.1 | |||
BC067095.1 | AAH67095.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |