Assay Details
Target Gene Details
Entrez Gene ID: | 256302 |
Gene Name: | N-acetyltransferase domain containing 1 |
Gene Aliases: |
C17orf103, Gtlf3b |
Location: |
Chr.17:21238870-21253266 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NATD1 | NM_152914.2 | NP_690878.2 | ||
AK126856.1 | ||||
BC029527.1 | AAH29527.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059146 | Chr.17:20849194 - 22546654 on Build GRCh38 | Gain | KCNJ18 TMEM11 MAP2K3 USP22 CCDC144NL-AS1 LOC105369205 LOC339260 CCDC144NL MTRNR2L1 NATD1 LINC01563 UBBP4 LOC100996792 FLJ36000 C17orf51 DHRS7B FAM27E5 KCNJ12 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |