Assay Details
Target Gene Details
Entrez Gene ID: | 443 |
Gene Name: | aspartoacylase |
Gene Aliases: |
ACY2, ASP |
Location: |
Chr.17:3474106-3502534 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASPA | NM_001128085.1 | NP_001121557.1 | ||
XM_017024661.1 | XP_016880150.1 | |||
AK312901.1 |
Target Gene Details
Entrez Gene ID: | 84690 |
Gene Name: | spermatogenesis associated 22 |
Gene Aliases: |
NYD-SP20, NYDSP20 |
Location: |
Chr.17:3440011-3513852 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPATA22 | NM_001321336.1 | NP_001308265.1 | ||
NM_001321337.1 | NP_001308266.1 | |||
AK295299.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892950 | Chr.17:3131812 - 3577153 on Build GRCh38 | Loss | OR1A1 OR1E1 TRPV1 OR3A3 ASPA OR1A2 TRPV3 OR3A2 OR3A4P OR1D4 SPATA22 OR1E2 OR3A1 |
nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain | OR1A1 CLUH LOC101927911 SHPK TRPV3 OR1G1 OR3A2 LOC105371591 NCBP3 RAP1GAP2 PAFAH1B1 CAMKK1 GSG2 OR1E2 P2RX5-TAX1BP3 OR3A1 LOC105371592 OR1E1 P2RX5 TAX1BP3 CCDC92B EMC6 TRPV1 OR3A3 ASPA OR1A2 MIR1253 P2RX1 OR3A4P OR1D5 ITGAE OR1D4 OR1D2 CTNS ATP2A3 SPATA22 MIR6776 |
nsv574232 | Chr.17:3254715 - 3503170 on Build GRCh38 | Gain | OR3A2 OR3A4P OR1E1 SPATA22 OR3A3 ASPA OR1E2 OR3A1 |
esv2660206 | Chr.17:2679367 - 3519614 on Build GRCh38 | Deletion | OR1A1 LOC105371592 OR1E1 CLUH LOC101927911 CCDC92B OR3A3 ASPA OR1A2 TRPV3 OR1G1 OR3A2 MIR1253 LOC105371591 OR3A4P OR1D5 OR1D4 RAP1GAP2 PAFAH1B1 OR1D2 SPATA22 MIR6776 OR1E2 OR3A1 |
nsv1055740 | Chr.17:3381841 - 3890153 on Build GRCh38 | Gain | OR1E1 P2RX5 TAX1BP3 EMC6 TRPV1 OR3A3 ASPA SHPK TRPV3 OR3A2 NCBP3 ITGAE CTNS SPATA22 CAMKK1 GSG2 OR1E2 P2RX5-TAX1BP3 |
nsv528368 | Chr.17:3459775 - 3507608 on Build GRCh38 | Gain | SPATA22 ASPA |
nsv833341 | Chr.17:3352243 - 3521390 on Build GRCh38 | Loss | TRPV3 OR3A2 OR1E1 SPATA22 OR3A3 ASPA OR1E2 |
nsv1060071 | Chr.17:3398474 - 3633472 on Build GRCh38 | Loss | TRPV3 TRPV1 SPATA22 OR3A3 ASPA OR1E2 SHPK |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |