Assay Details
Target Gene Details
Entrez Gene ID: | 5175 |
Gene Name: | platelet and endothelial cell adhesion molecule 1 |
Gene Aliases: |
CD31, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, endoCAM |
Location: |
Chr.17:64319415-64413844 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066968 | Chr.17:64215762 - 64332226 on Build GRCh38 | Gain | TEX2 PECAM1 |
nsv833513 | Chr.17:64235366 - 64396432 on Build GRCh38 | Loss | TEX2 PECAM1 |
nsv509666 | Chr.17:64254455 - 64396455 on Build GRCh38 | Insertion | TEX2 PECAM1 |
nsv833512 | Chr.17:64210750 - 64396432 on Build GRCh38 | Gain+Loss | TEX2 PECAM1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |