Assay Details
Target Gene Details
Entrez Gene ID: | 5892 |
Gene Name: | RAD51 paralog D |
Gene Aliases: |
BROVCA4, R51H3, RAD51L3, TRAD |
Location: |
Chr.17:35099792-35119869 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAD51D | NM_001142571.1 | NP_001136043.1 | ||
NM_002878.3 | NP_002869.3 | |||
NM_133629.2 | NP_598332.1 | |||
NR_037711.1 | ||||
NR_037712.1 | ||||
AB013341.1 | BAA25914.1 | |||
AB016223.1 | BAA31747.1 | |||
AB016224.1 | BAA31748.1 | |||
AB016225.1 | BAA31749.1 | |||
AB020412.1 | BAA34690.1 | |||
AF034956.1 | AAC39719.1 | |||
AK296241.1 | ||||
AK301249.1 | ||||
AL117459.1 | CAB55937.1 | |||
BC002723.2 | AAH02723.1 | |||
BC014422.1 | AAH14422.1 | |||
BX647297.1 | ||||
Y15572.1 | CAA75681.1 |
Target Gene Details
Entrez Gene ID: | 100529207 |
Gene Name: | RAD51L3-RFFL readthrough |
Gene Aliases: |
- |
Location: |
Chr.17:35009112-35121522 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAD51L3-RFFL | NR_037714.1 | |||
AK097811.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |