Assay Details
Target Gene Details
Entrez Gene ID: | 3927 |
Gene Name: | LIM and SH3 protein 1 |
Gene Aliases: |
Lasp-1, MLN50 |
Location: |
Chr.17:38869859-38921770 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LASP1 | NM_001271608.1 | NP_001258537.1 | ||
NM_006148.3 | NP_006139.1 | |||
NR_073384.1 | ||||
AK289847.1 | ||||
AK294704.1 | ||||
AK295522.1 | ||||
AK296092.1 | ||||
AK307868.1 | ||||
AK309933.1 | ||||
BC007560.1 | AAH07560.1 | |||
BC012460.1 | AAH12460.1 | |||
CN371189.1 | ||||
DC324561.1 | ||||
DC333179.1 | ||||
X82456.1 | CAA57833.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833439 | Chr.17:38743861 - 38923753 on Build GRCh38 | Loss | CWC25 PCGF2 MIR6779 LOC100287808 RPL23 LASP1 PIP4K2B MIR4727 SNORA21 C17orf98 PSMB3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |