Assay Details
Target Gene Details
Entrez Gene ID: | 1012 |
Gene Name: | cadherin 13 |
Gene Aliases: |
CDHH, P105 |
Location: |
Chr.16:82626794-83796610 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH13 | NM_001220488.1 | NP_001207417.1 | ||
NM_001220489.1 | NP_001207418.1 | |||
NM_001220490.1 | NP_001207419.1 | |||
NM_001220491.1 | NP_001207420.1 | |||
NM_001220492.1 | NP_001207421.1 | |||
NM_001257.4 | NP_001248.1 | |||
XM_017022848.1 | XP_016878337.1 | |||
XM_017022849.1 | XP_016878338.1 | |||
AK294277.1 | ||||
AK295886.1 | ||||
AK298612.1 | ||||
AK304592.1 | ||||
AK304733.1 | ||||
AK307862.1 | ||||
AU123178.1 | ||||
BC028624.2 | AAH28624.1 | |||
BC030653.2 | AAH30653.1 | |||
BI756586.1 | ||||
CV024366.1 | ||||
DC376661.1 | ||||
EU190357.1 | ||||
EU190358.1 | ||||
L34058.1 | AAA35624.1 | |||
U59288.1 | AAB18911.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv573371 | Chr.16:82131578 - 83629456 on Build GRCh38 | Loss | MIR8058 LOC101928417 MPHOSPH6 LOC101928446 CDH13 MIR3182 |
nsv573367 | Chr.16:82106956 - 83007241 on Build GRCh38 | Gain | MIR8058 LOC101928417 MPHOSPH6 LOC101928446 CDH13 |
nsv833305 | Chr.16:82530283 - 82688479 on Build GRCh38 | Gain | CDH13 |
nsv1061288 | Chr.16:82146152 - 83636829 on Build GRCh38 | Gain+Loss | MIR8058 LOC101928417 MPHOSPH6 LOC101928446 CDH13 MIR3182 |
dgv3051n100 | Chr.16:82138730 - 83654472 on Build GRCh38 | Gain | MIR8058 LOC101928417 MPHOSPH6 LOC101928446 CDH13 MIR3182 |
nsv1065896 | Chr.16:82179912 - 82698389 on Build GRCh38 | Gain | MIR8058 CDH13 |
dgv5253n54 | Chr.16:82148499 - 83630977 on Build GRCh38 | Gain | MIR8058 LOC101928417 MPHOSPH6 LOC101928446 CDH13 MIR3182 |
nsv1059718 | Chr.16:82530449 - 82776398 on Build GRCh38 | Loss | MIR8058 LOC101928446 CDH13 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |