Assay Details
Target Gene Details
Entrez Gene ID: | 2975 |
Gene Name: | general transcription factor IIIC subunit 1 |
Gene Aliases: |
TFIIIC, TFIIIC220, TFIIICalpha |
Location: |
Chr.16:27460613-27549930 on Build GRCh38 |
Assay Gene Location: | Within Intron 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTF3C1 | NM_001286242.1 | NP_001273171.1 | ||
NM_001520.3 | NP_001511.2 | |||
XM_017023188.1 | XP_016878677.1 | |||
BC044857.2 | AAH44857.1 | |||
BC137229.1 | ||||
BX648143.1 | CAH10771.1 | |||
CR627430.1 | CAH10517.1 | |||
U02619.1 | AAA17985.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059454 | Chr.16:27139937 - 27679877 on Build GRCh38 | Gain | NSMCE1 IL4R KIAA0556 GTF3C1 FLJ21408 LOC100128079 IL21R-AS1 IL21R KDM8 |
nsv833179 | Chr.16:27341363 - 27496185 on Build GRCh38 | Loss | IL4R GTF3C1 IL21R-AS1 IL21R |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |