Assay Details
Target Gene Details
Entrez Gene ID: | 23295 |
Gene Name: | mahogunin ring finger 1 |
Gene Aliases: |
RNF156 |
Location: |
Chr.16:4616493-4690974 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MGRN1 | XM_011522441.1 | XP_011520743.1 | ||
XM_017023093.1 | XP_016878582.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160333 | Chr.16:4570296 - 4709737 on Build GRCh38 | Deletion | UBALD1 ANKS3 MGRN1 NUDT16L1 C16orf96 MIR6769A |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | LOC105371063 EEF2KMT UBALD1 NAGPA-AS1 GLYR1 ZNF500 SMIM22 PPL CDIP1 C16orf71 MGRN1 ADCY9 CORO7 TFAP4 NUDT16L1 LINC01569 HMOX2 RBFOX1 SEPT12 NAGPA ROGDI PAM16 VASN GLIS2-AS1 ALG1 CORO7-PAM16 NMRAL1 ANKS3 GLIS2 DNAJA3 UBN1 C16orf89 C16orf96 MIR6769A SEC14L5 SRL |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |