Assay Details
Target Gene Details
Entrez Gene ID: | 1459 |
Gene Name: | casein kinase 2 alpha 2 |
Gene Aliases: |
CK2A2, CK2alpha', CSNK2A1 |
Location: |
Chr.16:58157907-58198116 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CSNK2A2 | NM_001896.2 | NP_001887.1 | ||
XM_005255800.3 | XP_005255857.1 | |||
XM_005255801.3 | XP_005255858.1 | |||
AI150700.1 | ||||
BC008812.2 | AAH08812.1 | |||
M55268.1 | AAA51548.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv572763 | Chr.16:58173299 - 58234532 on Build GRCh38 | Loss | CSNK2A2 CCDC113 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |