Assay Details
Target Gene Details
Entrez Gene ID: | 1800 |
Gene Name: | dipeptidase 1 (renal) |
Gene Aliases: |
MBD1, MDP, RDP |
Location: |
Chr.16:89613308-89641540 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPEP1 | NM_001128141.2 | NP_001121613.1 | ||
NM_004413.3 | NP_004404.1 | |||
XM_005256285.4 | XP_005256342.1 | |||
XM_005256286.3 | XP_005256343.1 | |||
XM_011522925.2 | XP_011521227.1 | |||
XM_017023009.1 | XP_016878498.1 | |||
AK309577.1 | ||||
BC017023.1 | AAH17023.1 | |||
BM152944.1 | ||||
BT006664.1 | AAP35310.1 | |||
CR457404.1 | CAG33685.1 | |||
D13138.1 | BAA02431.1 | |||
J05257.1 | AAB59410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv952078 | Chr.16:89602793 - 89640192 on Build GRCh38 | Deletion |
![]() |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv573739 | Chr.16:89625890 - 89696855 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv1063483 | Chr.16:89347250 - 89661187 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv509639 | Chr.16:89513538 - 89775915 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1066382 | Chr.16:89537178 - 89661187 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)