Assay Details
Target Gene Details
Entrez Gene ID: | 6376 |
Gene Name: | C-X3-C motif chemokine ligand 1 |
Gene Aliases: |
ABCD-3, C3Xkine, CXC3, CXC3C, NTN, NTT, SCYD1, fractalkine, neurotactin |
Location: |
Chr.16:57372461-57385048 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CX3CL1 | NM_001304392.1 | NP_001291321.1 | ||
NM_002996.4 | NP_002987.1 | |||
AB209037.1 | BAD92274.1 | |||
AK222692.1 | BAD96412.1 | |||
AK223351.1 | BAD97071.1 | |||
AK313806.1 | ||||
BC001163.1 | AAH01163.1 | |||
BC016164.1 | AAH16164.1 | |||
CA396869.1 | ||||
CR457426.1 | CAG33707.1 | |||
HY114023.1 | ||||
U84487.1 | AAB49679.1 | |||
U91835.1 | AAB50014.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |