Assay Details
Target Gene Details
Entrez Gene ID: | 1800 |
Gene Name: | dipeptidase 1 (renal) |
Gene Aliases: |
MBD1, MDP, RDP |
Location: |
Chr.16:89613308-89641540 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPEP1 | NM_001128141.2 | NP_001121613.1 | ||
XM_005256285.4 | XP_005256342.1 | |||
AK309577.1 | ||||
BM152944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | CHMP1A ZNF276 SPIRE2 CDK10 GAS8-AS1 CENPBD1 DBNDD1 GAS8 DEF8 MC1R TUBB3 LOC105371419 AFG3L1P SPATA33 TCF25 LOC107984817 VPS9D1 DPEP1 FANCA VPS9D1-AS1 SPATA2L |
nsv952078 | Chr.16:89602793 - 89640192 on Build GRCh38 | Deletion | DPEP1 |
nsv1929 | Chr.16:89601681 - 89628517 on Build GRCh38 | Insertion | DPEP1 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | CHMP1A PRDM7 ZNF276 SPIRE2 ZNF778 CDK10 GAS8-AS1 LOC105371414 ANKRD11 URAHP GAS8 MC1R LOC101927817 TUBB3 LOC105371419 SPG7 DPEP1 TUBB8P7 LOC100287036 VPS9D1-AS1 SNORD68 FAM157C RPL13 CENPBD1 DBNDD1 DEF8 AFG3L1P SPATA33 TCF25 LOC107984817 VPS9D1 FANCA SPATA2L CPNE7 |
nsv573738 | Chr.16:89549357 - 89625890 on Build GRCh38 | Loss | SNORD68 SPG7 RPL13 DPEP1 CPNE7 |
nsv1063483 | Chr.16:89347250 - 89661187 on Build GRCh38 | Gain | CHMP1A LOC101927817 SNORD68 SPG7 SPATA33 RPL13 DPEP1 ANKRD11 CPNE7 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | ZNF276 CDH15 SNAI3-AS1 PABPN1L CDT1 LOC105371414 ANKRD11 URAHP SNAI3 GAS8 LOC339059 MC1R LOC105371409 LOC101927817 TUBB3 DPEP1 TUBB8P7 VPS9D1-AS1 PIEZO1 RNF166 SNORD68 IL17C FAM157C RPL13 CYBA GALNS CBFA2T3 TCF25 LOC107984817 FANCA SPATA2L CPNE7 CHMP1A PRDM7 TRAPPC2L SPIRE2 LOC107987238 ACSF3 ZNF778 CDK10 GAS8-AS1 LOC100289580 CTU2 LINC00304 SLC22A31 MVD LOC105371419 SPG7 LOC100287036 LOC100129697 LOC400558 CENPBD1 DBNDD1 MIR4722 DEF8 LOC101927793 AFG3L1P SPATA33 APRT VPS9D1 |
nsv509639 | Chr.16:89513538 - 89775915 on Build GRCh38 | Insertion | CHMP1A ZNF276 SNORD68 RPL13 CDK10 SPG7 SPATA33 VPS9D1 DPEP1 FANCA VPS9D1-AS1 SPATA2L CPNE7 |
nsv1066382 | Chr.16:89537178 - 89661187 on Build GRCh38 | Gain | CHMP1A SNORD68 SPG7 SPATA33 RPL13 DPEP1 CPNE7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |