Assay Details
Target Gene Details
Entrez Gene ID: | 1428 |
Gene Name: | crystallin mu |
Gene Aliases: |
DFNA40, THBP |
Location: |
Chr.16:21258518-21303136 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRYM | NM_001888.4 | NP_001879.1 | ||
AK290852.1 | ||||
AK310424.1 | ||||
BC018061.1 | AAH18061.1 | |||
BI860684.1 | ||||
BX648477.1 | CAI46030.1 | |||
L02950.1 | AAC16914.1 | |||
U85772.1 | AAB81564.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv571623 | Chr.16:21084049 - 21341186 on Build GRCh38 | Gain | DNAH3 TMEM159 ANKS4B ZP2 CRYM-AS1 CRYM |
esv3638164 | Chr.16:21185596 - 21338776 on Build GRCh38 | Gain | ANKS4B ZP2 CRYM-AS1 CRYM |
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | GPR139 PDILT KNOP1 TMC5 C16orf62 CLEC19A LOC100190986 TMEM159 LOC81691 NPIPB3 CRYM-AS1 TMC7 GP2 LYRM1 SNX29P1 DNAH3 ACSM2B GPRC5B ERI2 ANKS4B IQCK SYT17 ACSM1 COQ7 DCUN1D3 GDE1 ACSM2A CRYM ITPRIPL2 ACSM5 ACSM3 SMG1P3 CCP110 ZP2 THUMPD1 UMOD LOC102723385 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |