Assay Details
Target Gene Details
Entrez Gene ID: | 221188 |
Gene Name: | adhesion G protein-coupled receptor G5 |
Gene Aliases: |
GPR114, PGR27 |
Location: |
Chr.16:57529339-57578182 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRG5 | XM_011522949.2 | XP_011521251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833245 | Chr.16:57434073 - 57584619 on Build GRCh38 | Loss | POLR2C CIAPIN1 CCDC102A DOK4 COQ9 ADGRG5 |
nsv1055537 | Chr.16:57531862 - 57570612 on Build GRCh38 | Loss | CCDC102A ADGRG5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |