Assay Details
Target Gene Details
Entrez Gene ID: | 23199 |
Gene Name: | Gse1 coiled-coil protein |
Gene Aliases: |
CRHSP24, KIAA0182 |
Location: |
Chr.16:85169512-85676206 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSE1 | XM_005255859.4 | XP_005255916.3 | ||
XM_005255860.3 | XP_005255917.3 | |||
XM_005255861.4 | XP_005255918.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3639458 | Chr.16:85192805 - 85201062 on Build GRCh38 | Loss | GSE1 |
esv2664171 | Chr.16:85192756 - 85201071 on Build GRCh38 | Deletion | GSE1 |
nsv833312 | Chr.16:85158268 - 85327826 on Build GRCh38 | Loss | GSE1 MIR5093 LINC00311 |
dgv3059n100 | Chr.16:85068826 - 85219999 on Build GRCh38 | Gain | GSE1 FAM92B KIAA0513 LOC400548 |
nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain | LOC146513 LOC101928474 LINC00917 KCNG4 MIR1910 CRISPLD2 EMC8 ZDHHC7 LINC00311 TAF1C FAM92B LOC101928614 FLJ30679 GSE1 FOXF1 COX4I1 TLDC1 LINC01081 GINS2 ATP2C2 LOC400548 IRF8 FOXC2 ADAD2 FENDRR MIR6774 LOC105376777 LOC105371374 FOXC2-AS1 MIR5093 MTHFSD COTL1 LOC102724344 LINC01082 C16orf74 KIAA0513 FOXL1 WFDC1 USP10 LOC440390 KLHL36 LOC101928557 DNAAF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |