Assay Details
Target Gene Details
Entrez Gene ID: | 1800 |
Gene Name: | dipeptidase 1 (renal) |
Gene Aliases: |
MBD1, MDP, RDP |
Location: |
Chr.16:89613308-89641540 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPEP1 | NM_001128141.2 | NP_001121613.1 | ||
NM_004413.3 | NP_004404.1 | |||
XM_005256285.4 | XP_005256342.1 | |||
XM_005256286.3 | XP_005256343.1 | |||
AK309577.1 | ||||
BM152944.1 | ||||
J05257.1 | AAB59410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1057351 | Chr.16:89610380 - 89707988 on Build GRCh38 | Loss | VPS9D1 CDK10 SPATA33 SPATA2L DPEP1 CHMP1A |
nsv952078 | Chr.16:89602793 - 89640192 on Build GRCh38 | Deletion | DPEP1 |
nsv1929 | Chr.16:89601681 - 89628517 on Build GRCh38 | Insertion | DPEP1 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | SNORD68 AFG3L1P ANKRD11 CENPBD1 SPIRE2 TCF25 CHMP1A GAS8-AS1 MC1R VPS9D1 ZNF778 ZNF276 SPATA33 URAHP TUBB3 LOC105371414 VPS9D1-AS1 RPL13 SPATA2L LOC105371419 LOC100287036 DBNDD1 SPG7 DEF8 CDK10 FANCA LOC101927817 FAM157C DPEP1 CPNE7 GAS8 LOC107984817 TUBB8P7 PRDM7 |
nsv573738 | Chr.16:89549357 - 89625890 on Build GRCh38 | Loss | SPG7 SNORD68 RPL13 DPEP1 CPNE7 |
nsv1063483 | Chr.16:89347250 - 89661187 on Build GRCh38 | Gain | SPG7 SNORD68 RPL13 SPATA33 ANKRD11 LOC101927817 DPEP1 CPNE7 CHMP1A |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | IL17C SNAI3 CDT1 SNAI3-AS1 CENPBD1 SPIRE2 CHMP1A GAS8-AS1 ACSF3 MC1R LOC400558 ZNF276 SPATA33 TUBB3 LOC105371414 LOC105371409 SPATA2L LOC100287036 CBFA2T3 DBNDD1 LOC107987238 SPG7 MVD PABPN1L LOC100289580 DPEP1 SLC22A31 CTU2 LOC101927793 SNORD68 AFG3L1P ANKRD11 LINC00304 TCF25 VPS9D1 LOC100129697 GALNS ZNF778 APRT LOC339059 URAHP CYBA RNF166 MIR4722 VPS9D1-AS1 TRAPPC2L RPL13 LOC105371419 PIEZO1 DEF8 CDK10 FANCA CDH15 LOC101927817 FAM157C CPNE7 GAS8 LOC107984817 TUBB8P7 PRDM7 |
nsv509639 | Chr.16:89513538 - 89775915 on Build GRCh38 | Insertion | SNORD68 RPL13 SPATA2L CHMP1A SPG7 VPS9D1 ZNF276 CDK10 FANCA SPATA33 DPEP1 CPNE7 VPS9D1-AS1 |
nsv1066382 | Chr.16:89537178 - 89661187 on Build GRCh38 | Gain | SPG7 SNORD68 RPL13 SPATA33 DPEP1 CPNE7 CHMP1A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |