Assay Details
Target Gene Details
Entrez Gene ID: | 146198 |
Gene Name: | ZFP90 zinc finger protein |
Gene Aliases: |
FIK, NK10, ZNF756, zfp-90 |
Location: |
Chr.16:68533484-68576072 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZFP90 | NM_001305203.1 | NP_001292132.1 | ||
NM_001305204.1 | NP_001292133.1 | |||
NM_001305206.1 | NP_001292135.1 | |||
NM_001305207.1 | NP_001292136.1 | |||
NM_001305208.1 | NP_001292137.1 | |||
NM_133458.3 | NP_597715.2 | |||
NR_130976.1 | ||||
NR_130977.1 | ||||
NR_130978.1 | ||||
XM_005255804.2 | XP_005255861.1 | |||
XM_017022952.1 | XP_016878441.1 | |||
AK074332.1 | ||||
AK304152.1 | ||||
AK316251.1 | ||||
BC140886.1 | ||||
BC140887.1 | ||||
DA803284.1 | ||||
KC249978.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115264806] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |