Assay Details
Target Gene Details
Entrez Gene ID: | 1071 |
Gene Name: | cholesteryl ester transfer protein |
Gene Aliases: |
BPIFF, HDLCQ10 |
Location: |
Chr.16:56961923-56983844 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CETP | NM_000078.2 | NP_000069.2 | ||
NM_001286085.1 | NP_001273014.1 | |||
XM_006721124.3 | XP_006721187.1 | |||
AK291586.1 | ||||
AK297699.1 | ||||
BC025739.1 | AAH25739.1 | |||
BT020064.1 | AAV38867.1 | |||
KU177972.1 | ||||
KU177973.1 | ||||
M30185.1 | AAA51977.1 | |||
M83573.1 | AAB59388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059098 | Chr.16:56733938 - 57028336 on Build GRCh38 | Gain | MIR6863 HERPUD1 MIR138-2 CETP SLC12A3 NUP93 NLRC5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |