Assay Details
Target Gene Details
Entrez Gene ID: | 1800 |
Gene Name: | dipeptidase 1 (renal) |
Gene Aliases: |
MBD1, MDP, RDP |
Location: |
Chr.16:89613308-89641540 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPEP1 | NM_001128141.2 | NP_001121613.1 | ||
NM_004413.3 | NP_004404.1 | |||
XM_005256285.4 | XP_005256342.1 | |||
XM_005256286.3 | XP_005256343.1 | |||
XM_011522925.2 | XP_011521227.1 | |||
XM_017023009.1 | XP_016878498.1 | |||
AK309577.1 | ||||
BM152944.1 | ||||
J05257.1 | AAB59410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | TUBB3 LOC107984817 FANCA VPS9D1 DPEP1 DBNDD1 CDK10 ZNF276 SPATA33 CHMP1A TCF25 MC1R SPATA2L AFG3L1P VPS9D1-AS1 GAS8 SPIRE2 CENPBD1 DEF8 GAS8-AS1 LOC105371419 |
nsv952078 | Chr.16:89602793 - 89640192 on Build GRCh38 | Deletion | DPEP1 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | TUBB3 FANCA DBNDD1 ZNF276 RPL13 LOC100287036 MC1R AFG3L1P VPS9D1-AS1 ANKRD11 SNORD68 TUBB8P7 CENPBD1 GAS8-AS1 LOC101927817 SPG7 LOC107984817 PRDM7 VPS9D1 URAHP DPEP1 CDK10 SPATA33 CHMP1A TCF25 LOC105371414 SPATA2L GAS8 FAM157C ZNF778 SPIRE2 CPNE7 DEF8 LOC105371419 |
nsv573739 | Chr.16:89625890 - 89696855 on Build GRCh38 | Gain | DPEP1 CDK10 SPATA33 CHMP1A SPATA2L |
nsv1063483 | Chr.16:89347250 - 89661187 on Build GRCh38 | Gain | ANKRD11 SNORD68 CPNE7 DPEP1 SPATA33 RPL13 CHMP1A LOC101927817 SPG7 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | LOC105371409 FANCA CTU2 DBNDD1 CDH15 SNAI3-AS1 RPL13 LOC339059 LOC101927793 AFG3L1P VPS9D1-AS1 SNORD68 TUBB8P7 PABPN1L LOC101927817 SLC22A31 LOC107984817 LOC400558 APRT URAHP PIEZO1 DPEP1 MVD FAM157C ZNF778 SPIRE2 DEF8 CDT1 TUBB3 CYBA ZNF276 CBFA2T3 LOC100287036 MC1R ANKRD11 RNF166 CENPBD1 LOC100289580 GAS8-AS1 LOC100129697 LOC107987238 SPG7 MIR4722 TRAPPC2L PRDM7 VPS9D1 CDK10 SPATA33 CHMP1A GALNS TCF25 ACSF3 LOC105371414 SNAI3 SPATA2L IL17C LINC00304 GAS8 CPNE7 LOC105371419 |
nsv509639 | Chr.16:89513538 - 89775915 on Build GRCh38 | Insertion | FANCA VPS9D1 DPEP1 CDK10 ZNF276 SPATA33 RPL13 CHMP1A SPATA2L VPS9D1-AS1 SNORD68 CPNE7 SPG7 |
nsv1066382 | Chr.16:89537178 - 89661187 on Build GRCh38 | Gain | SNORD68 CPNE7 DPEP1 SPATA33 RPL13 CHMP1A SPG7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |